Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCTGCTGTGCAAAAAGAAAGAC[A/G]GTACCAAGAAGGGACGTGTTAATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608255 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAF3IP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRAF3IP3 - TRAF3 interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287754.1 | 121 | Intron | NP_001274683.1 | |||
NM_001320143.1 | 121 | Intron | NP_001307072.1 | |||
NM_001320144.1 | 121 | UTR 5 | NP_001307073.1 | |||
NM_025228.3 | 121 | UTR 5 | NP_079504.2 | |||
XM_005273279.4 | 121 | Intron | XP_005273336.1 | |||
XM_005273280.2 | 121 | Intron | XP_005273337.1 | |||
XM_011510018.2 | 121 | UTR 5 | XP_011508320.1 | |||
XM_011510019.1 | 121 | UTR 5 | XP_011508321.1 | |||
XM_017002399.1 | 121 | UTR 5 | XP_016857888.1 | |||
XM_017002400.1 | 121 | UTR 5 | XP_016857889.1 |