Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCCTTGTCCAGCTTCAGTTTTTC[A/G]ATTTCACGCTTAATTGGGGCAAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604733 | ||||||||||||||||||||
Literature Links: |
WARS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WARS2 - tryptophanyl tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015836.3 | 813 | Silent Mutation | ATC,ATT | I,I 321 | NP_056651.1 | |
NM_201263.2 | 813 | UTR 3 | NP_957715.1 | |||
XM_005270350.3 | 813 | Silent Mutation | ATC,ATT | I,I 303 | XP_005270407.1 | |
XM_011540494.2 | 813 | Silent Mutation | ATC,ATT | I,I 298 | XP_011538796.1 | |
XM_011540495.2 | 813 | Silent Mutation | ATC,ATT | I,I 235 | XP_011538797.1 | |
XM_017000038.1 | 813 | Silent Mutation | ATC,ATT | I,I 302 | XP_016855527.1 | |
XM_017000039.1 | 813 | Silent Mutation | ATC,ATT | I,I 298 | XP_016855528.1 | |
XM_017000040.1 | 813 | Silent Mutation | ATC,ATT | I,I 264 | XP_016855529.1 | |
XM_017000041.1 | 813 | Silent Mutation | ATC,ATT | I,I 208 | XP_016855530.1 | |
XM_017000042.1 | 813 | UTR 3 | XP_016855531.1 |