Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGTTGTCATGGCGACCAGAAAG[C/G]ACACAGAGGAGGAGCAGGTAGTCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602171 | ||||||||||||||||||||
Literature Links: |
DUSP27 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DUSP27 - dual specificity phosphatase 27 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080426.1 | 179 | Missense Mutation | CAC,GAC | H,D 6 | NP_001073895.1 | |
XM_005245603.3 | 179 | Missense Mutation | CAC,GAC | H,D 6 | XP_005245660.1 | |
XM_011510146.2 | 179 | Intron | XP_011508448.1 |
GPA33 - glycoprotein A33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105371598 - uncharacterized LOC105371598 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |