Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTAAAAATTTTAGCCAATGGCC[G/T]CCTTCAGTTGGGACATGGTCTTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604774 MIM: 615730 | ||||||||||||||||||||
Literature Links: |
ANGPTL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANGPTL3 - angiopoietin like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014495.3 | 229 | Missense Mutation | CGC,CTC | R,L 50 | NP_055310.1 |
DOCK7 - dedicator of cytokinesis 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271999.1 | 229 | Intron | NP_001258928.1 | |||
NM_001272000.1 | 229 | Intron | NP_001258929.1 | |||
NM_001272001.1 | 229 | Intron | NP_001258930.1 | |||
NM_001272002.1 | 229 | Intron | NP_001258931.1 | |||
NM_033407.3 | 229 | Intron | NP_212132.2 | |||
XM_005271292.2 | 229 | Intron | XP_005271349.1 | |||
XM_011542326.2 | 229 | Intron | XP_011540628.1 | |||
XM_011542327.2 | 229 | Intron | XP_011540629.1 | |||
XM_011542328.2 | 229 | Intron | XP_011540630.1 | |||
XM_011542330.2 | 229 | Intron | XP_011540632.1 | |||
XM_017002639.1 | 229 | Intron | XP_016858128.1 | |||
XM_017002640.1 | 229 | Intron | XP_016858129.1 |