Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCCCACTCCAAACAGCCCGGTAA[A/C]ACGGTATGTTTCTCCTGCCACCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608335 | ||||||||||||||||||||
Literature Links: |
LOC105371433 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105371433 - uncharacterized LOC105371433 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLEKHO1 - pleckstrin homology domain containing O1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304722.1 | 596 | UTR 5 | NP_001291651.1 | |||
NM_001304723.1 | 596 | UTR 5 | NP_001291652.1 | |||
NM_001304724.1 | 596 | UTR 5 | NP_001291653.1 | |||
NM_016274.5 | 596 | Silent Mutation | AAA,AAC | K,N 105 | NP_057358.2 | |
XM_017001420.1 | 596 | UTR 5 | XP_016856909.1 |