Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCCCTCACCTCTCTCGCTCAGC[A/G]GCCAAGGTGGGCGTCTCCACCACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615679 MIM: 609151 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP85 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP85 - centrosomal protein 85 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH3BGRL3 - SH3 domain binding glutamate rich protein like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBXN11 - UBX domain protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077262.1 | 925 | Silent Mutation | GCC,GCT | A,A 259 | NP_001070730.1 | |
NM_145345.2 | 925 | Silent Mutation | GCC,GCT | A,A 346 | NP_663320.2 | |
NM_183008.2 | 925 | Silent Mutation | GCC,GCT | A,A 379 | NP_892120.2 |