Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGGACTGAAGACACTCAATTTG[A/G]GAAGCTGTGTGATCGCCACAAACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605619 | ||||||||||||||||||||
Literature Links: |
IL20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL20 - interleukin 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018724.3 | 561 | Missense Mutation | AGA,GGA | R,G 31 | NP_061194.2 | |
XM_005273152.3 | 561 | Missense Mutation | AGA,GGA | R,G 116 | XP_005273209.1 |