Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGAGCAACTGGTGTGTCGTTTA[C/T]CGAAAACGGATATTTTCATTATATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 613975 | ||||||||||||||||||||
Literature Links: |
IFI44L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IFI44L - interferon induced protein 44 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006820.3 | 387 | Missense Mutation | CCG,TCG | P,S 117 | NP_006811.2 | |
XM_005270391.3 | 387 | Intron | XP_005270448.1 | |||
XM_005270392.4 | 387 | Intron | XP_005270449.1 | |||
XM_005270393.4 | 387 | Intron | XP_005270450.1 | |||
XM_006710304.3 | 387 | Intron | XP_006710367.1 | |||
XM_011540539.2 | 387 | Missense Mutation | CCG,TCG | P,S 117 | XP_011538841.1 | |
XM_017000120.1 | 387 | Intron | XP_016855609.1 |