Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACGAAGGCTCCAATCCGCCGGCT[A/T]CCCCACTGCAAGGTGACCCCAGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 153390 | ||||||||||||||||||||
Literature Links: |
LCK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LCK - LCK proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042771.2 | 294 | Missense Mutation | ACC,TCC | T,S 59 | NP_001036236.1 | |
NM_005356.4 | 294 | Missense Mutation | ACC,TCC | T,S 59 | NP_005347.3 | |
XM_011541453.2 | 294 | Missense Mutation | ACC,TCC | T,S 59 | XP_011539755.1 | |
XM_011541454.2 | 294 | Missense Mutation | ACC,TCC | T,S 59 | XP_011539756.1 | |
XM_011541455.2 | 294 | Missense Mutation | ACC,TCC | T,S 59 | XP_011539757.1 |