Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGGCTGTGCAGGGCTTCTACCC[C/T]CTCCACCGACAGCAGCAGATCACGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601738 MIM: 609617 | ||||||||||||||||||||
Literature Links: |
EXTL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EXTL1 - exostosin like glycosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC30A2 - solute carrier family 30 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004434.2 | 1137 | Missense Mutation | NP_001004434.1 | |||
NM_032513.4 | 1137 | Missense Mutation | NP_115902.1 |