Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGTGACCAGGGCACCATAGGTCA[A/G]GGTGAAGAGCTCAGAGCTCTAAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610955 | ||||||||||||||||||||
Literature Links: |
TRAPPC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRAPPC3 - trafficking protein particle complex 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270894.1 | 197 | Silent Mutation | CTG,TTG | L,L 21 | NP_001257823.1 | |
NM_001270895.1 | 197 | UTR 5 | NP_001257824.1 | |||
NM_001270896.1 | 197 | UTR 5 | NP_001257825.1 | |||
NM_001270897.1 | 197 | Intron | NP_001257826.1 | |||
NM_014408.4 | 197 | Silent Mutation | CTG,TTG | L,L 21 | NP_055223.1 |