Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACGGTACATGAGCAGTAGGATCAG[C/G]AAGACGGCAAAGAGGATGCCCACGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616104 MIM: 600017 | ||||||||||||||||||||
Literature Links: |
RBPJL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RBPJL - recombination signal binding protein for immunoglobulin kappa J region like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SDC4 - syndecan 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002999.3 | 384 | Missense Mutation | TTC,TTG | F,L 163 | NP_002990.2 | |
XM_011528977.2 | 384 | Missense Mutation | TTC,TTG | F,L 91 | XP_011527279.1 |