Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGGAATGATCGCTAACGAGCAGG[A/G]TCCTGGCATGATTTAGAACGGGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603307 MIM: 609114 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BFSP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BFSP1 - beaded filament structural protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DSTN - destrin, actin depolymerizing factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011546.1 | Intron | NP_001011546.1 | ||||
NM_006870.3 | Intron | NP_006861.1 | ||||
XM_011529142.1 | Intron | XP_011527444.1 | ||||
XM_011529143.1 | Intron | XP_011527445.1 | ||||
XM_011529144.1 | Intron | XP_011527446.1 |