Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCACCGACATGAAGGGAACAGGC[A/G]TCATGGACTGTGCGCCCAAGGTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600369 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASS4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CASS4 - Cas scaffolding protein family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164114.1 | 803 | Missense Mutation | ATC,GTC | I,V 6 | NP_001157586.1 | |
NM_001164115.1 | 803 | Missense Mutation | ATC,GTC | I,V 6 | NP_001157587.1 | |
NM_001164116.1 | 803 | Missense Mutation | ATC,GTC | I,V 6 | NP_001157588.1 | |
NM_020356.3 | 803 | Missense Mutation | ATC,GTC | I,V 6 | NP_065089.2 | |
XM_006723831.3 | 803 | Missense Mutation | ATC,GTC | I,V 6 | XP_006723894.1 |
CSTF1 - cleavage stimulation factor subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |