Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTCTTTCATGTTTAACCTGAAA[C/T]GAACGTAACTATAAATGAGTATCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611386 MIM: 603503 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADNP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADNP - activity dependent neuroprotector homeobox | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282531.1 | 854 | Intron | NP_001269460.1 | |||
NM_001282532.1 | 854 | Intron | NP_001269461.1 | |||
NM_015339.3 | 854 | Intron | NP_056154.1 | |||
NM_181442.2 | 854 | Intron | NP_852107.1 | |||
XM_011528747.2 | 854 | Intron | XP_011527049.1 | |||
XM_011528748.2 | 854 | Intron | XP_011527050.1 | |||
XM_017027757.1 | 854 | Intron | XP_016883246.1 | |||
XM_017027758.1 | 854 | Intron | XP_016883247.1 | |||
XM_017027759.1 | 854 | Intron | XP_016883248.1 |
ADNP-AS1 - ADNP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DPM1 - dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317034.1 | 854 | UTR 3 | NP_001303963.1 | |||
NM_001317035.1 | 854 | UTR 3 | NP_001303964.1 | |||
NM_001317036.1 | 854 | UTR 3 | NP_001303965.1 | |||
NM_003859.2 | 854 | UTR 3 | NP_003850.1 | |||
XM_011529093.2 | 854 | Intron | XP_011527395.1 |