Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCATCATCTGGCGGGCAGCCCCC[C/T]GATTTCTCTGTAAGAGAATCTCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602905 MIM: 605161 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNS1 - potassium voltage-gated channel modifier subfamily S member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WFDC5 - WAP four-disulfide core domain 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145652.3 | 182 | Silent Mutation | TCA,TCG | S,S 31 | NP_663627.1 | |
XM_011528601.1 | 182 | Silent Mutation | TCA,TCG | S,S 31 | XP_011526903.1 | |
XM_011528602.1 | 182 | Silent Mutation | TCA,TCG | S,S 31 | XP_011526904.1 |