Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGTGCTCCCTATCCAGTCCAAA[C/G]GACCCTCTTGATTACAGCACAGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606577 | ||||||||||||||||||||
Literature Links: |
C20orf24 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C20orf24 - chromosome 20 open reading frame 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199534.1 | 623 | UTR 3 | NP_001186463.1 | |||
NM_018840.4 | 623 | UTR 3 | NP_061328.1 | |||
NM_199483.2 | 623 | Missense Mutation | ACG,AGG | T,R 100 | NP_955777.1 |
SLA2 - Src like adaptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032214.3 | 623 | Intron | NP_115590.1 | |||
NM_175077.2 | 623 | Intron | NP_778252.1 | |||
XM_017028098.1 | 623 | Intron | XP_016883587.1 |
TGIF2-C20orf24 - TGIF2-C20orf24 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199535.1 | 623 | UTR 3 | NP_001186464.1 |