Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTGGCTGAATTCTGGGCCCTCAC[A/C]AGCCTCAAGCGGCGCCACCAGAACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STK35 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STK35 - serine/threonine kinase 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080836.3 | 4603 | Silent Mutation | ACA,ACC | T,T 252 | NP_543026.2 | |
XM_011529174.2 | 4603 | Silent Mutation | ACA,ACC | T,T 348 | XP_011527476.1 |