Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAAACTTTTGTCTTCTCCGGCTG[C/T]AATGGCAACCTTAACAACTTCAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SPINT4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPINT4 - serine peptidase inhibitor, Kunitz type 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178455.1 | 241 | Silent Mutation | TGC,TGT | C,C 74 | NP_848550.1 |
WFDC3 - WAP four-disulfide core domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080614.1 | 241 | Intron | NP_542181.1 | |||
XM_011528553.2 | 241 | Intron | XP_011526855.1 | |||
XM_017027668.1 | 241 | Intron | XP_016883157.1 |