Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTGGTGGGACAAGACAACGGGC[C/T]GGTGGAGGTGTCCACATTGCAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPEPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NPEPL1 - aminopeptidase-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204872.1 | 523 | Missense Mutation | CCG,CTG | P,L 134 | NP_001191801.1 | |
NM_001204873.1 | 523 | Missense Mutation | CCG,CTG | P,L 114 | NP_001191802.1 | |
NM_024663.3 | 523 | Missense Mutation | CCG,CTG | P,L 162 | NP_078939.3 |
STX16-NPEPL1 - STX16-NPEPL1 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |