Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCACCCTCTCTCTCCACAGGCC[C/T]GCAGGTCACACTCTGCCACAGCGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C20orf141 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C20orf141 - chromosome 20 open reading frame 141 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256538.1 | 438 | Silent Mutation | CCC,CCT | P,P 88 | NP_001243467.1 | |
NM_080739.2 | 438 | Silent Mutation | CCC,CCT | P,P 88 | NP_542777.1 |
TMEM239 - transmembrane protein 239 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167670.2 | 438 | Intron | NP_001161142.1 | |||
NM_001318207.1 | 438 | Intron | NP_001305136.1 | |||
XM_011529128.2 | 438 | Intron | XP_011527430.1 | |||
XM_011529129.2 | 438 | Intron | XP_011527431.1 |