Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGGGGTTCAGGCACGAGTTGGC[A/G]TAGCTGAGGCTGGTGATGACGTAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600731 MIM: 602548 | ||||||||||||||||||||
Literature Links: |
NPBWR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NPBWR2 - neuropeptides B/W receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005286.2 | 918 | Silent Mutation | TAC,TAT | Y,Y 306 | NP_005277.2 |
OPRL1 - opioid related nociceptin receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |