Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAGTCGAGTCCTCACTCTTTTCT[C/G]TTCTAAGACATGCATCTTTTTCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616621 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX27 - DEAD-box helicase 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNFX1 - zinc finger NFX1-type containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021035.2 | 4352 | Missense Mutation | CAG,GAG | Q,E 1727 | NP_066363.1 | |
XM_006723836.1 | 4352 | Missense Mutation | CAG,GAG | Q,E 1727 | XP_006723899.1 | |
XM_011528924.1 | 4352 | Missense Mutation | CAG,GAG | Q,E 1677 | XP_011527226.1 | |
XM_017027971.1 | 4352 | Missense Mutation | CAG,GAG | Q,E 1009 | XP_016883460.1 |