Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCCCTCAGCCCTTCGGGCAGTTC[C/T]GGGAAGAGGTAGAGCCTGGAGTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614694 MIM: 614425 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPRD1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPRD1B - regulation of nuclear pre-mRNA domain containing 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021215.3 | Intron | NP_067038.1 | ||||
XM_005260477.3 | Intron | XP_005260534.1 | ||||
XM_005260480.4 | Intron | XP_005260537.1 | ||||
XM_011528954.2 | Intron | XP_011527256.1 | ||||
XM_017027990.1 | Intron | XP_016883479.1 | ||||
XM_017027991.1 | Intron | XP_016883480.1 |
TTI1 - TELO2 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |