Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTGAATGTGAATCCAGCCCTGC[C/T]CTTGAGTCCCACAGGTCTTGCAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607412 | ||||||||||||||||||||
Literature Links: |
BPIFA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BPIFA1 - BPI fold containing family A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243193.1 | 192 | Missense Mutation | CCC,CTC | P,L 41 | NP_001230122.1 | |
NM_016583.3 | 192 | Missense Mutation | CCC,CTC | P,L 41 | NP_057667.1 | |
NM_130852.2 | 192 | Missense Mutation | CCC,CTC | P,L 41 | NP_570913.1 |