Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCCCAGGGCTTTGTATCCCCCA[C/T]CCGAAGGCTGGTGTTCCCCAAGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCM2L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCM2L - CCM2 like scaffolding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080625.3 | 123 | Missense Mutation | ACC,ATC | T,I 16 | NP_542192.2 | |
XM_006723706.3 | 123 | Missense Mutation | ACC,ATC | T,I 16 | XP_006723769.1 | |
XM_011528566.1 | 123 | Missense Mutation | ACC,ATC | T,I 16 | XP_011526868.1 | |
XM_011528567.2 | 123 | Missense Mutation | ACC,ATC | T,I 9 | XP_011526869.1 | |
XM_011528568.2 | 123 | Missense Mutation | ACC,ATC | T,I 8 | XP_011526870.1 | |
XM_011528569.1 | 123 | Missense Mutation | ACC,ATC | T,I 16 | XP_011526871.1 |