Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGAAACTGCACAAAGAATGGAA[A/T]AGTGAAGGAAGCCCAGGTAAGAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611120 | ||||||||||||||||||||
Literature Links: |
SPTLC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPTLC3 - serine palmitoyltransferase long chain base subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018327.2 | 158 | Missense Mutation | AAA,ATA | K,I 34 | NP_060797.2 | |
XM_011529279.1 | 158 | Missense Mutation | AAA,ATA | K,I 34 | XP_011527581.1 | |
XM_011529280.2 | 158 | Intron | XP_011527582.1 | |||
XM_017027922.1 | 158 | Missense Mutation | AAA,ATA | K,I 34 | XP_016883411.1 |