Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCATTTCAACTGCTCTCTGTCAC[C/T]TTGCATGGTAGCCTTTTTTTTGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611386 | ||||||||||||||||||||
Literature Links: |
ADNP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADNP - activity dependent neuroprotector homeobox | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282531.1 | 4296 | Missense Mutation | AGT,GGT | S,G 1021 | NP_001269460.1 | |
NM_001282532.1 | 4296 | Missense Mutation | AGT,GGT | S,G 1021 | NP_001269461.1 | |
NM_015339.3 | 4296 | Missense Mutation | AGT,GGT | S,G 1021 | NP_056154.1 | |
NM_181442.2 | 4296 | Missense Mutation | AGT,GGT | S,G 1021 | NP_852107.1 | |
XM_011528747.2 | 4296 | Missense Mutation | AGT,GGT | S,G 1021 | XP_011527049.1 | |
XM_011528748.2 | 4296 | Missense Mutation | AGT,GGT | S,G 1030 | XP_011527050.1 | |
XM_017027757.1 | 4296 | Missense Mutation | AGT,GGT | S,G 1021 | XP_016883246.1 | |
XM_017027758.1 | 4296 | Missense Mutation | AGT,GGT | S,G 1021 | XP_016883247.1 | |
XM_017027759.1 | 4296 | Missense Mutation | AGT,GGT | S,G 1021 | XP_016883248.1 |