Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTCATATGTCTGAATGCTCTC[C/T]GTGGAAATCTTCTCGATAGATTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603307 MIM: 162151 | ||||||||||||||||||||
Literature Links: |
BFSP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BFSP1 - beaded filament structural protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161705.1 | 1939 | Silent Mutation | ACA,ACG | T,T 508 | NP_001155177.1 | |
NM_001195.4 | 1939 | Silent Mutation | ACA,ACG | T,T 633 | NP_001186.1 | |
NM_001278606.1 | 1939 | Silent Mutation | ACA,ACG | T,T 494 | NP_001265535.1 | |
NM_001278607.1 | 1939 | Silent Mutation | ACA,ACG | T,T 522 | NP_001265536.1 | |
NM_001278608.1 | 1939 | Silent Mutation | ACA,ACG | T,T 494 | NP_001265537.1 | |
XM_017028005.1 | 1939 | Silent Mutation | ACA,ACG | T,T 597 | XP_016883494.1 |
PCSK2 - proprotein convertase subtilisin/kexin type 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |