Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGCTCTTCCACAGGAATGTTGA[C/G]GATGACATCCATGTCTGGGGTGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C20orf96 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C20orf96 - chromosome 20 open reading frame 96 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080571.1 | 1155 | Missense Mutation | CTC,GTC | L,V 352 | NP_542138.1 | |
NM_153269.2 | 1155 | Missense Mutation | CTC,GTC | L,V 353 | NP_695001.2 | |
XM_006723543.3 | 1155 | Missense Mutation | CTC,GTC | L,V 300 | XP_006723606.1 |