Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCCCAGGAGGAGAGCTACGAGG[A/T]GTGGAGCAGTAGGAGCCAGCGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603889 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIRPB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SIRPB1 - signal regulatory protein beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005260641.3 | 1494 | Missense Mutation | CAC,CTC | H,L 370 | XP_005260698.1 | |
XM_005260643.3 | 1494 | Missense Mutation | CAC,CTC | H,L 259 | XP_005260700.1 | |
XM_005260644.3 | 1494 | Missense Mutation | CAC,CTC | H,L 153 | XP_005260701.1 | |
XM_011529130.2 | 1494 | Intron | XP_011527432.1 | |||
XM_011529134.2 | 1494 | Intron | XP_011527436.1 | |||
XM_017027577.1 | 1494 | Missense Mutation | CAC,CTC | H,L 371 | XP_016883066.1 | |
XM_017027578.1 | 1494 | Intron | XP_016883067.1 | |||
XM_017027579.1 | 1494 | Intron | XP_016883068.1 | |||
XM_017027580.1 | 1494 | Intron | XP_016883069.1 | |||
XM_017027581.1 | 1494 | Silent Mutation | ACA,ACT | T,T 327 | XP_016883070.1 | |
XM_017027582.1 | 1494 | Silent Mutation | ACA,ACT | T,T 327 | XP_016883071.1 | |
XM_017027583.1 | 1494 | Intron | XP_016883072.1 | |||
XM_017027584.1 | 1494 | Intron | XP_016883073.1 | |||
XM_017027585.1 | 1494 | Missense Mutation | CAC,CTC | H,L 260 | XP_016883074.1 |
SIRPD - signal regulatory protein delta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |