Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCATCACTCCTAAAATCTTCAAT[A/G]TGTGTTTCCAAAAGCCAAGGATGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615757 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIZ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIZ - kizuna centrosomal protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163022.1 | Intron | NP_001156494.1 | ||||
NM_001163023.1 | Intron | NP_001156495.1 | ||||
NM_001276389.1 | Intron | NP_001263318.1 | ||||
NM_018474.4 | Intron | NP_060944.3 | ||||
XM_011529296.2 | Intron | XP_011527598.1 | ||||
XM_011529297.2 | Intron | XP_011527599.1 | ||||
XM_011529298.2 | Intron | XP_011527600.1 | ||||
XM_011529299.2 | Intron | XP_011527601.1 | ||||
XM_017027951.1 | Intron | XP_016883440.1 | ||||
XM_017027952.1 | Intron | XP_016883441.1 | ||||
XM_017027953.1 | Intron | XP_016883442.1 | ||||
XM_017027954.1 | Intron | XP_016883443.1 |