Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGGCTTGGAGCTCTTCCACCTC[A/G]GCCCTTAATACAGGCATGTTCTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C20orf96 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C20orf96 - chromosome 20 open reading frame 96 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080571.1 | 1052 | Silent Mutation | GCC,GCT | A,A 317 | NP_542138.1 | |
NM_153269.2 | 1052 | Silent Mutation | GCC,GCT | A,A 318 | NP_695001.2 | |
XM_006723543.3 | 1052 | Silent Mutation | GCC,GCT | A,A 265 | XP_006723606.1 |