Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGCTACGGGCTACGAACCTGAAA[A/G]GGCAGGAAGGAGAGCGCGTGCTCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611203 MIM: 610866 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC5 - DnaJ heat shock protein family (Hsp40) member C5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1914 - microRNA 1914 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR647 - microRNA 647 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UCKL1 - uridine-cytidine kinase 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193379.1 | 1391 | Silent Mutation | CCC,CCT | P,P 355 | NP_001180308.1 | |
NM_017859.3 | 1391 | Silent Mutation | CCC,CCT | P,P 370 | NP_060329.2 | |
XM_005260216.2 | 1391 | Silent Mutation | CCC,CCT | P,P 378 | XP_005260273.1 | |
XM_006723806.2 | 1391 | Silent Mutation | CCC,CCT | P,P 380 | XP_006723869.1 | |
XM_006723807.2 | 1391 | Silent Mutation | CCC,CCT | P,P 379 | XP_006723870.1 | |
XM_006723808.1 | 1391 | Silent Mutation | CCC,CCT | P,P 379 | XP_006723871.1 | |
XM_006723809.1 | 1391 | Silent Mutation | CCC,CCT | P,P 372 | XP_006723872.1 | |
XM_006723810.1 | 1391 | Silent Mutation | CCC,CCT | P,P 371 | XP_006723873.1 | |
XM_006723811.3 | 1391 | Silent Mutation | CCC,CCT | P,P 364 | XP_006723874.1 | |
XM_011528868.2 | 1391 | Silent Mutation | CCC,CCT | P,P 365 | XP_011527170.1 | |
XM_011528869.1 | 1391 | Silent Mutation | CCC,CCT | P,P 264 | XP_011527171.1 | |
XM_011528870.1 | 1391 | Silent Mutation | CCC,CCT | P,P 254 | XP_011527172.1 | |
XM_011528871.1 | 1391 | Silent Mutation | CCC,CCT | P,P 254 | XP_011527173.1 | |
XM_011528872.1 | 1391 | Silent Mutation | CCC,CCT | P,P 254 | XP_011527174.1 | |
XM_011528873.2 | 1391 | Intron | XP_011527175.1 | |||
XM_017027894.1 | 1391 | Intron | XP_016883383.1 | |||
XM_017027895.1 | 1391 | Intron | XP_016883384.1 | |||
XM_017027896.1 | 1391 | Silent Mutation | CCC,CCT | P,P 371 | XP_016883385.1 | |
XM_017027897.1 | 1391 | Intron | XP_016883386.1 | |||
XM_017027898.1 | 1391 | Silent Mutation | CCC,CCT | P,P 370 | XP_016883387.1 | |
XM_017027899.1 | 1391 | Silent Mutation | CCC,CCT | P,P 369 | XP_016883388.1 | |
XM_017027900.1 | 1391 | Silent Mutation | CCC,CCT | P,P 355 | XP_016883389.1 | |
XM_017027901.1 | 1391 | Silent Mutation | CCC,CCT | P,P 354 | XP_016883390.1 | |
XM_017027902.1 | 1391 | Silent Mutation | CCC,CCT | P,P 256 | XP_016883391.1 | |
XM_017027903.1 | 1391 | Silent Mutation | CCC,CCT | P,P 252 | XP_016883392.1 | |
XM_017027904.1 | 1391 | Silent Mutation | CCC,CCT | P,P 164 | XP_016883393.1 | |
XM_017027905.1 | 1391 | Silent Mutation | CCC,CCT | P,P 162 | XP_016883394.1 | |
XM_017027906.1 | 1391 | Silent Mutation | CCC,CCT | P,P 162 | XP_016883395.1 | |
XM_017027907.1 | 1391 | Silent Mutation | CCC,CCT | P,P 151 | XP_016883396.1 | |
XM_017027908.1 | 1391 | Silent Mutation | CCC,CCT | P,P 151 | XP_016883397.1 |