Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATATAGTTTAATAGGAAATACTT[A/C]TTTTAAAAGACTGACCCCTTTGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604205 MIM: 607179 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPNE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPNE1 - copine 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198863.1 | 4731 | Intron | NP_001185792.1 | |||
NM_003915.5 | 4731 | Intron | NP_003906.2 | |||
NM_152925.2 | 4731 | Intron | NP_690902.1 | |||
NM_152926.2 | 4731 | Intron | NP_690903.1 | |||
NM_152927.2 | 4731 | Intron | NP_690904.1 | |||
NM_152928.2 | 4731 | Intron | NP_690905.1 |
RBM12 - RNA binding motif protein 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198838.1 | 4731 | UTR 3 | NP_001185767.1 | |||
NM_001198840.1 | 4731 | UTR 3 | NP_001185769.1 | |||
NM_006047.5 | 4731 | UTR 3 | NP_006038.2 | |||
NM_152838.3 | 4731 | UTR 3 | NP_690051.1 |