Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATATCTATGCAAACACTTATGTGA[A/G]TCTCACCGAGATTGTCAAGCAAATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
WFDC10A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WFDC10A - WAP four-disulfide core domain 10A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080753.2 | 147 | Intron | NP_542791.1 | |||
XM_017027679.1 | 147 | Silent Mutation | GAA,GAG | E,E 49 | XP_016883168.1 |
WFDC9 - WAP four-disulfide core domain 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_147198.3 | 147 | Intron | NP_671731.1 |