Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCTGCTCCCGCACGTTGTTGGT[A/G]GTGTTGTCCTCAGAGGCTGAGTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601920 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JAG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JAG1 - jagged 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000214.2 | 3849 | Silent Mutation | ACC,ACT | T,T 1111 | NP_000205.1 | |
XM_017027707.1 | 3849 | Silent Mutation | ACC,ACT | T,T 952 | XP_016883196.1 |
MIR6870 - microRNA 6870 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |