Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCATTGTGTCTGCAGCCCGTGGT[C/G]GGGGGAAGATGTCCCCCTACACCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607976 | ||||||||||||||||||||
Literature Links: |
COX4I2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COX4I2 - cytochrome c oxidase subunit 4I2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032609.2 | 166 | Missense Mutation | CGG,GGG | R,G 31 | NP_115998.2 | |
XM_005260579.4 | 166 | Missense Mutation | CGG,GGG | R,G 36 | XP_005260636.1 | |
XM_005260580.4 | 166 | Missense Mutation | CGG,GGG | R,G 36 | XP_005260637.1 | |
XM_005260581.3 | 166 | Missense Mutation | CGG,GGG | R,G 31 | XP_005260638.1 |