Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGCAATTAAATGGCCTTTCCCCC[A/G]AGTGTGTCCACTGATGTCTAAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF337 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF337 - zinc finger protein 337 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290261.1 | 1571 | Missense Mutation | TCG,TTG | S,L 567 | NP_001277190.1 | |
NM_015655.3 | 1571 | Missense Mutation | TCG,TTG | S,L 567 | NP_056470.1 | |
XM_005260702.3 | 1571 | Missense Mutation | TCG,TTG | S,L 535 | XP_005260759.1 | |
XM_006723558.3 | 1571 | Missense Mutation | TCG,TTG | S,L 567 | XP_006723621.1 | |
XM_011529219.2 | 1571 | Missense Mutation | TCG,TTG | S,L 567 | XP_011527521.1 | |
XM_017027802.1 | 1571 | Missense Mutation | TCG,TTG | S,L 535 | XP_016883291.1 | |
XM_017027803.1 | 1571 | Missense Mutation | TCG,TTG | S,L 535 | XP_016883292.1 | |
XM_017027804.1 | 1571 | Missense Mutation | TCG,TTG | S,L 502 | XP_016883293.1 |
ZNF337-AS1 - ZNF337 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |