Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTACTGCATGTGCTGCAGCGACTG[A/C]GCGCTGTAGGCAGAAAAGGGAATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604612 | ||||||||||||||||||||
Literature Links: |
NKX2-2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NKX2-2 - NK2 homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002509.3 | 731 | Silent Mutation | GCG,GCT | A,A 239 | NP_002500.1 | |
XM_006723566.3 | 731 | Silent Mutation | GCG,GCT | A,A 168 | XP_006723629.1 |