Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGGAGGCCGTCTGTCCAGTTTG[C/T]ATGTGTGGATCAGTCTCTCTGAGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608550 | ||||||||||||||||||||
Literature Links: |
PCED1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCED1A - PC-esterase domain containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271168.1 | 1609 | Missense Mutation | TAC,TGC | Y,C 388 | NP_001258097.1 | |
NM_022760.4 | 1609 | Missense Mutation | TAC,TGC | Y,C 439 | NP_073597.2 | |
XM_005260804.2 | 1609 | Missense Mutation | TAC,TGC | Y,C 439 | XP_005260861.1 | |
XM_005260805.2 | 1609 | Missense Mutation | TAC,TGC | Y,C 388 | XP_005260862.1 | |
XM_005260806.3 | 1609 | Missense Mutation | TAC,TGC | Y,C 347 | XP_005260863.1 |
VPS16 - VPS16, CORVET/HOPS core subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |