Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCAGGCAAGAAACAGCCAGGGA[A/T]GAAACCTATCAAGGGCATCACCAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614108 MIM: 614110 | ||||||||||||||||||||
Literature Links: |
BPIFB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BPIFB2 - BPI fold containing family B member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BPIFB6 - BPI fold containing family B member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174897.2 | 173 | Missense Mutation | AAG,ATG | K,M 58 | NP_777557.1 | |
XM_017027663.1 | 173 | Missense Mutation | AAG,ATG | K,M 74 | XP_016883152.1 |