Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACGTCAGTGATTCCTGGCCTGAA[C/G]AACATCATTGAGTGAGTTGTCCTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607412 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BPIFA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BPIFA1 - BPI fold containing family A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243193.1 | 379 | Missense Mutation | AAC,AAG | N,K 103 | NP_001230122.1 | |
NM_016583.3 | 379 | Missense Mutation | AAC,AAG | N,K 103 | NP_057667.1 | |
NM_130852.2 | 379 | Missense Mutation | AAC,AAG | N,K 103 | NP_570913.1 |