Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCAGGGTCTCGCTCTGTCGACT[G/T]TCTTAAGAAAGGGCTCTGTCCAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615076 MIM: 605937 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGME1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MGME1 - mitochondrial genome maintenance exonuclease 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001310338.1 | Intron | NP_001297267.1 | ||||
NM_001310339.1 | Intron | NP_001297268.1 | ||||
NM_052865.3 | Intron | NP_443097.1 | ||||
XM_005260867.2 | Intron | XP_005260924.1 | ||||
XM_005260870.3 | Intron | XP_005260927.1 | ||||
XM_006723663.3 | Intron | XP_006723726.1 | ||||
XM_011529394.1 | Intron | XP_011527696.1 | ||||
XM_011529395.1 | Intron | XP_011527697.1 | ||||
XM_017028127.1 | Intron | XP_016883616.1 | ||||
XM_017028128.1 | Intron | XP_016883617.1 |
SNORD17 - small nucleolar RNA, C/D box 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNX5 - sorting nexin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282454.1 | Intron | NP_001269383.1 | ||||
NM_014426.3 | Intron | NP_055241.1 | ||||
NM_152227.2 | Intron | NP_689413.1 |