Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCATCAGAAGAATATATACATGGA[G/T]CGTGTTTGGGGAGGAGATGGAGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602968 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCAS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCAS1 - breast carcinoma amplified sequence 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316361.2 | 1909 | UTR 3 | NP_001303290.1 | |||
NM_001323347.1 | 1909 | UTR 3 | NP_001310276.1 | |||
NM_003657.3 | 1909 | UTR 3 | NP_003648.2 | |||
XM_005260587.2 | 1909 | UTR 3 | XP_005260644.1 | |||
XM_005260589.2 | 1909 | UTR 3 | XP_005260646.1 | |||
XM_005260590.2 | 1909 | UTR 3 | XP_005260647.1 | |||
XM_005260591.2 | 1909 | UTR 3 | XP_005260648.1 | |||
XM_005260593.2 | 1909 | UTR 3 | XP_005260650.1 | |||
XM_005260594.2 | 1909 | UTR 3 | XP_005260651.1 | |||
XM_005260595.2 | 1909 | UTR 3 | XP_005260652.1 | |||
XM_011529090.2 | 1909 | UTR 3 | XP_011527392.1 | |||
XM_011529091.2 | 1909 | UTR 3 | XP_011527393.1 | |||
XM_017028110.1 | 1909 | UTR 3 | XP_016883599.1 | |||
XM_017028111.1 | 1909 | UTR 3 | XP_016883600.1 | |||
XM_017028112.1 | 1909 | UTR 3 | XP_016883601.1 |