Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTGGGATTCATGGAGGAGGCTG[C/T]AAAGAGTGTGGAGACTTTGGTCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608712 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPRT - protein tyrosine phosphatase, receptor type T | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007050.5 | 12277 | UTR 3 | NP_008981.4 | |||
NM_133170.3 | 12277 | UTR 3 | NP_573400.3 | |||
XM_017027611.1 | 12277 | UTR 3 | XP_016883100.1 | |||
XM_017027612.1 | 12277 | UTR 3 | XP_016883101.1 | |||
XM_017027613.1 | 12277 | UTR 3 | XP_016883102.1 |