Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATGACCCACGGCCGCATGGTGC[C/T]GAGAGGGACAGGCTTCTCCGGAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614824 | ||||||||||||||||||||
Literature Links: |
AP5S1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AP5S1 - adaptor related protein complex 5 sigma 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204446.1 | 380 | Silent Mutation | GCC,GCT | A,A 43 | NP_001191375.1 | |
NM_001204447.1 | 380 | Silent Mutation | GCC,GCT | A,A 43 | NP_001191376.1 | |
NM_018347.2 | 380 | Silent Mutation | GCC,GCT | A,A 43 | NP_060817.1 |
LOC101929125 - uncharacterized LOC101929125 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |