Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACAGTTCCATGTCTTGGCTAACA[C/T]GGCTCTCACCGCTGGCCTCAACACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615884 MIM: 603253 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APMAP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APMAP - adipocyte plasma membrane associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020531.2 | 1857 | UTR 3 | NP_065392.1 | |||
XM_005260763.3 | 1857 | UTR 3 | XP_005260820.1 |
CST7 - cystatin F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |