Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTGCAATAGAACACACAATCTTT[G/T]GTGAGCACATCTTGTGAATATATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614118 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSHZ2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSHZ2 - teashirt zinc finger homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193421.1 | Intron | NP_001180350.1 | ||||
NM_173485.5 | Intron | NP_775756.3 | ||||
XM_017027640.1 | Intron | XP_016883129.1 | ||||
XM_017027641.1 | Intron | XP_016883130.1 |