Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGGGAAGGGAGGCTGGGCTAGA[A/T]GGGGTCACAAGGCCTGTGTAGGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605832 MIM: 612342 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACSS2 - acyl-CoA synthetase short-chain family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001076552.2 | 20 | Intron | NP_001070020.2 | |||
NM_001242393.1 | 20 | UTR 5 | NP_001229322.1 | |||
NM_018677.3 | 20 | Intron | NP_061147.1 | |||
XM_005260455.1 | 20 | Intron | XP_005260512.1 | |||
XM_005260456.1 | 20 | Intron | XP_005260513.1 | |||
XM_006723826.1 | 20 | UTR 5 | XP_006723889.1 | |||
XM_011528905.1 | 20 | Intron | XP_011527207.1 | |||
XM_011528906.1 | 20 | Intron | XP_011527208.1 | |||
XM_011528907.1 | 20 | Intron | XP_011527209.1 | |||
XM_011528908.1 | 20 | Intron | XP_011527210.1 | |||
XM_011528909.2 | 20 | Intron | XP_011527211.1 | |||
XM_011528911.1 | 20 | UTR 5 | XP_011527213.1 | |||
XM_011528912.2 | 20 | Intron | XP_011527214.1 |
GGT7 - gamma-glutamyltransferase 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |